Pyridinium chloride

Warning

Synonyms: Pyridine hydrochloride, Pyridinehydrochloride, Pyridine HCl

243086-100G 243086-5G 243086-500G
220047-299EA 56.75 USD
220047-299 220047-301 220047-300
Pyridinium chloride
Pyridinium chloride
Formula: C₅H₆ClN
MW: 115.56 g/mol
Boiling Pt: 222.5 °C (1013 hPa)
Melting Pt: 144 °C
Density: 1.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012802
CAS Number: 628-13-7
EINECS: 211-027-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR