Sodium diphenylamine-4-sulfonate ACS

Synonyms: Sodium Diphenylaminesulphonate, Sodium N-phenylsulphanilate, Sodium (4-anilinobenzenesulphonate), Diphenylamine-4-sulphonic acid, sodium salt

242969-25G 242969-5G
220047-288EA 463.08 USD
220047-288 220047-289
Sodium diphenylamine-4-sulfonate ACS
Sodium diphenylamine-4-sulphonate
Formula: C₁₂H₁₀NNaO₃S
MW: 271.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00036468
CAS Number: 6152-67-6
EINECS: 228-165-6
Merck Index: 13,07397

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR