4-Bromo-N,N-dimethylaniline ≥97%

242950-100G 242950-25G
220047-286EA 134.62 USD
220047-286 220047-287
4-Bromo-N,N-dimethylaniline ≥97%
4-Bromo-N,N-dimethylaniline
Formula: C₈H₁₀BrN
MW: 200.08 g/mol
Boiling Pt: 264 °C (1013 hPa)
Melting Pt: 54…55 °C
Density: 1.3218 g/cm³ (99.5 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000093
CAS Number: 586-77-6
EINECS: 209-582-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR