Dimethyl fumarate ≥97%

Synonyms: Fumaric acid dimethyl ester

242926-25G 242926-100G
220047-282EA 34.78 USD
220047-282 220047-281
Dimethyl fumarate ≥97%
Dimethyl fumarate
Formula: C₆H₈O₄
MW: 144.13 g/mol
Boiling Pt: 192 °C (1013 hPa)
Melting Pt: 102 °C
Density: 1.37 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064438
CAS Number: 624-49-7
EINECS: 210-849-0
Merck Index: 13,04308

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR