1-Bromo-2-iodobenzene ≥99%

Warning

Synonyms: 2-Bromo-1-iodobenzene, 2-Bromoiodobenzene

242616-5G 242616-25G
220047-249EA 46.85 USD
220047-249 220047-248
1-Bromo-2-iodobenzene ≥99%
1-Bromo-2-iodobenzene
Formula: C₆H₄BrI
MW: 282.91 g/mol
Boiling Pt: 250 °C (1013 hPa)
Density: 2.2571 g/cm³ (25 °C)
MDL Number: MFCD00001030
CAS Number: 583-55-1
EINECS: 209-508-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR