sec-Butyl acetate ≥99%

Danger

Synonyms: 1-Methylpropyl acetate

242594-5ML 242594-100ML
220047-246EA 41.46 USD
220047-246 220047-245
sec-Butyl acetate ≥99%
sec-Butyl acetate
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 112 °C (1013 hPa)
Melting Pt: -98.9 °C
Density: 0.87 g/cm³ (20 °C)
Flash Pt: 19 °C
MDL Number: MFCD00009328
CAS Number: 105-46-4
EINECS: 203-300-1
UN: 1123
ADR: 3,II
Merck Index: 13,01535

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR