Benzoic acid ≥99.5% ACS

Warning

242381-500G 242381-100G 242381-25G 242381-3KG
220047-224EA 69.15 USD
220047-224 220047-221 220047-222 220047-223
Benzoic acid ≥99.5% ACS
Benzoic acid
Formula: C₇H₆O₂
MW: 122.12 g/mol
Boiling Pt: 249 °C (1013 hPa)
Melting Pt: 121…123 °C
Density: 1.316 g/cm³ (20 °C)
Flash Pt: 121 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002398
CAS Number: 65-85-0
EINECS: 200-618-2
REACH: 01-2119455536-33
Merck Index: 13,01092
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d561f311b11ea8ab2c8f07253e6748e588eb71aa&vid=dcf5e45072cda5106216b497cd29716270ae772c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR