2-Chloroethyl methyl ether ≥98%

Synonyms: 2-Methoxyethyl chloride

242349-100G 242349-5G
220047-215EA 161.54 USD
220047-215 220047-216
2-Chloroethyl methyl ether ≥98%
2-Chloroethyl methyl ether
Formula: C₃H₇ClO
MW: 94.54 g/mol
Boiling Pt: 89…90 °C (1013 hPa)
Melting Pt: –55…55 °C
Density: 1.04 g/cm³ (20 °C)
MDL Number: MFCD00000972
CAS Number: 627-42-9
EINECS: 210-999-7
UN: 3271
ADR: 3,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR