(R)-(-)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol

Synonyms: (R)-(-)-1,2-O-Isopropylidenglycerol, (R)-(-)-2,2-Dimethyl-1,3-dioxolane-4-methanol, (R)-(-)-2,3-O-Isopropylideneglycerol, (R)-(-)-1,2-O-Isopropylideneglycerol, (R)-(2,2-Dimethyl-1,3-dioxolan-4-yl)methanol

241806-5G 241806-1G
220047-174EA 76.14 USD
220047-174 220047-173
(R)-(-)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol
(R)-(-)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol
Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Boiling Pt: 72 °C (1013 hPa)
Density: 1.07 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00003213
CAS Number: 14347-78-5
EINECS: 405-320-8
Merck Index: 12,05232

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR