Phenyl vinyl sulfone ≥99%

Synonyms: Vinyl phenyl sulphone, (Vinylsulphonyl)benzene

241717-1G 241717-5G
220046-890EA 48.89 USD
220046-890 220046-891
Phenyl vinyl sulfone ≥99%
Phenyl vinyl sulphone
Formula: C₈H₈O₂S
MW: 168.22 g/mol
MDL Number: MFCD00007554
CAS Number: 5535-48-8
EINECS: 226-890-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR