1,2-Bis(2-chloroethoxy)ethane ≥97%

241628-100G 241628-500G
220046-878EA 75.6 USD
220046-878 220046-879
1,2-Bis(2-chloroethoxy)ethane ≥97%
1,2-Bis(2-chloroethoxy)ethane
Formula: ClH₂CCH₂OCH₂CH₂OCH₂CH₂Cl
MW: 187.07 g/mol
Boiling Pt: 235 °C (1013 hPa)
Melting Pt: -31.5 °C
Density: 1.1024 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000976
CAS Number: 112-26-5
EINECS: 203-952-7
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR