Lead(II,IV) oxide ≥99%

Danger

Synonyms: Minium, Orange lead

241547-100G 241547-500G
220047-155EA 62.77 USD
220047-155 220047-156
Lead(II,IV) oxide ≥99%
Lead(II,IV) oxide
Formula: Pb₃O₄
MW: 685.6 g/mol
Boiling Pt: 2682 °C (1013 hPa)
Melting Pt: 830 °C
Density: 9.1 g/cm³ (20 °C)
MDL Number: MFCD00078491
CAS Number: 1314-41-6
EINECS: 215-235-6
UN: 1479
ADR: 5.1,II
Merck Index: 12,05449

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR