Silver (I) sulfide

Warning

Synonyms: Disilver sulphide

241474-5G 241474-25G
220047-152EA 145.23 USD
220047-152 220047-151
Silver (I) sulfide
Silver(I) sulphide
Formula: Ag₂S
MW: 247.8 g/mol
Melting Pt: 842 °C
Density: 7.23 g/cm³ (25 °C)
MDL Number: MFCD00003406
CAS Number: 21548-73-2
EINECS: 244-438-2
Merck Index: 12,08677

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR