Chromium hexacarbonyl

Synonyms: Chromium carbonyl, Hexacarbonylchromium, Hexacarbonylchromium(0), Chromium(0) hexacarbonyl

241458-50G 241458-10G
220047-150EA 355.85 USD
220047-150 220047-149
Chromium hexacarbonyl
Chromium hexacarbonyl
Formula: C₆CrO₆
MW: 220.06 g/mol
Boiling Pt: 210 °C (1013 hPa)
Melting Pt: 90 °C
Density: 1.77 g/cm³ (20 °C)
MDL Number: MFCD00010945
CAS Number: 13007-92-6
EINECS: 235-852-4
UN: 3466
ADR: 6.1,II
Merck Index: 13,02253

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR