5-Chloroquinoline

Warning

24098-1G-F 24098-U
220046-770EA 25.08 USD
220046-770 220046-771
5-Chloroquinoline
5-Chloroquinoline
Formula: C₉H₆ClN
MW: 163.61 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00239413
CAS Number: 635-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR