Trimethyl phosphite ≥99%

Synonyms: TMP

240907-25G 240907-100G
220045-393EA 29.55 USD
220045-393 220045-392
Trimethyl phosphite ≥99%
Trimethyl phosphite
Formula: C₃H₉O₃P
MW: 124.08 g/mol
Boiling Pt: 111 °C (1013 hPa)
Melting Pt: –77.8 °C
Density: 1.042 g/cm³ (20 °C)
Flash Pt: 50 °C
MDL Number: MFCD00008350
CAS Number: 121-45-9
EINECS: 204-471-5
UN: 2329
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR