Succinic anhydride

Warning

Synonyms: Dihydrofuran-2,5-dione

239690-250G 239690-50G
220046-260EA 234.08 USD
220046-260 220046-261
Succinic anhydride
Succinic anhydride
Formula: C₄H₄O₃
MW: 100.07 g/mol
Boiling Pt: 261 °C (1013 hPa)
Melting Pt: 120 °C
Density: 1.503 g/cm³ (20 °C)
Flash Pt: 157 °C
MDL Number: MFCD00005525
CAS Number: 108-30-5
EINECS: 203-570-0
UN: 3261
ADR: 8,III
Merck Index: 12,09039

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR