4,4'-Isopropylidenediphenol ≥99% analytical standard, for drug analysis

Synonyms: Bisphenol A, BPA, 2,2-Bis(4-hydroxyphenyl)propane

239658-250G 239658-50G
220046-258EA 507.69 USD
220046-258 220046-259
4,4'-Isopropylidenediphenol ≥99% analytical standard, for drug analysis
4,4'-Isopropylidenediphenol
Formula: C₁₅H₁₆O₂
MW: 228.29 g/mol
Boiling Pt: 401 °C (1013 hPa)
Melting Pt: 155 °C
Density: 1.205 g/cm³ (20 °C)
Flash Pt: 227 °C
MDL Number: MFCD00002366
CAS Number: 80-05-7
EINECS: 201-245-8
Merck Index: 13,01296
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63888e215660c5847e91f6773838f54f30e7191f&vid=63888e215660c5847e91f6773838f54f30e7191f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR