meta-Phosphoric acid ACS

Danger

Synonyms: Metaphosphoric acid

239275-2.5KG 239275-5G
220046-852EA 1114.62 USD
220046-852 220046-854 220046-853 220046-851
meta-Phosphoric acid ACS
meta-Phosphoric acid
Formula: (HPO₃)n
MW: 79.98 g/mol
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: 21 °C
Density: 2.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011351
CAS Number: 37267-86-0
EINECS: 253-433-4
UN: 3260
ADR: 8,III

Order Now

Specification Test Results

Assay 33.5-36.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR