Chromium(III) nitrate nonahydrate

Warning

Synonyms: Chromium trinitrate nonahydrate, Chromic nitrate nonahydrate, Chromium nitrate nonahydrate

239259-2KG 239259-100G 239259-500G
220046-846EA 309.54 USD
220046-846 220046-845 220046-847
Chromium(III) nitrate nonahydrate
Chromium(III) nitrate nonahydrate
Formula: CrN₃O₉·9H₂O
MW: 400.15 g/mol
Melting Pt: 60 °C
Density: 1.807 g/cm³ (20 °C)
MDL Number: MFCD00149659
CAS Number: 7789-02-8
EINECS: 236-921-1
UN: 2720
ADR: 5.1,III
Merck Index: 13,02246

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR