Aluminium hydroxide, Reagent Grade

Warning

239186-25G 239186-500G 239186-2KG
220046-757EA 33.42 USD
220046-757 220046-759 220046-758
Aluminium hydroxide, Reagent Grade
Aluminium hydroxide
Formula: Al(OH)₃
Melting Pt: 300 °C
Density: 2.42 g/cm³ (20 °C)
MDL Number: MFCD00003420
CAS Number: 21645-51-2
EINECS: 244-492-7
Merck Index: 12,00355

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR