Ammonium trifluoroacetate ≥98%

Synonyms: Ammoniumtrifluoroacetate, Trifluoroacetic acid ammonium salt

238732-100G 238732-25G
220046-719EA 198.69 USD
220046-719 220046-720
Ammonium trifluoroacetate ≥98%
Ammonium trifluoroacetate
Formula: C₂H₄F₃NO₂
MW: 131.05 g/mol
MDL Number: MFCD00012615
CAS Number: 3336-58-1
EINECS: 222-072-4

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR