4-Methoxyphenyl isocyanate ≥99%

Synonyms: 1-Isocyanato-4-methoxybenzene

238600-5G 238600-25G
220046-704EA 63 USD
220046-704 220046-703
4-Methoxyphenyl isocyanate ≥99%
4-Methoxyphenyl isocyanate
Formula: C₈H₇NO₂
MW: 149.15 g/mol
Boiling Pt: 106 °C (1013 hPa)
Density: 1.1559 g/cm³ (20 °C)
MDL Number: MFCD00002026
CAS Number: 5416-93-3
EINECS: 226-513-1
UN: 2206
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR