Cerium trinitrate hexahydrate ≥99%

Danger

Synonyms: Cerous chloride hexahydrate, Cerium (III) nitrate hexahydrate

238538-100G 238538-500G
220046-689EA 127.38 USD
220046-689 220046-690
Cerium trinitrate hexahydrate ≥99%
Cerium trinitrate hexahydrate
Formula: Ce(NO₃)₃·6H₂O
MW: 434.22 g/mol
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: 39 °C
Density: 2.377 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149631
CAS Number: 10294-41-4
EINECS: 233-297-2
UN: 1477
ADR: 5.1,II

Order Now

Specification Test Results

Assay 99% trace metals basis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR