1-Bromo-2-propanol ≥70%, Technical Grade

Danger

238430-50G 238430-10G
220045-381EA 247.69 USD
220045-381 220045-380
1-Bromo-2-propanol ≥70%, Technical Grade
1-Bromo-2-propanol
Formula: BrH₂CCH(OH)CH₃
MW: 138.99 g/mol
Boiling Pt: 145 °C (1013 hPa)
Melting Pt: -55 °C
Density: 1.53 g/cm³ (20 °C)
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00004529
CAS Number: 19686-73-8
EINECS: 243-225-1
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR