1,3-Diiodopropane ≥99%

Warning

Synonyms: Trimethylene diiodide

238414-100G 238414-25G
220045-376EA 515.85 USD
220045-376 220045-377
1,3-Diiodopropane ≥99%
1,3-Diiodopropane
Formula: C₃H₆I₂
MW: 295.89 g/mol
Boiling Pt: 228 °C (1013 hPa)
Melting Pt: –20 °C
Density: 2.59617 g/cm³ (4 °C)
MDL Number: MFCD00001097
CAS Number: 627-31-6
EINECS: 210-993-4

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR