Ethyl methyl sulfide ≥96%

238317-100G 238317-25G
220046-671EA 161.54 USD
220046-671 220046-672
Ethyl methyl sulfide ≥96%
Ethyl methyl sulphide
Formula: C₃H₈S
MW: 76.16 g/mol
Boiling Pt: 65 °C (1013 hPa)
Melting Pt: –104.8 °C
Density: 0.837 g/cm³ (20 °C)
MDL Number: MFCD00009268
CAS Number: 624-89-5
EINECS: 210-868-4
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR