Dimethyl maleate ≥96%

Warning

Synonyms: Dimethyl Maleate, Maleic acid dimethyl ester

238198-100G 238198-1KG
220046-648EA 34.68 USD
220046-648 220046-649
Dimethyl maleate ≥96%
Dimethyl maleate
Formula: C₆H₈O₄
MW: 144.13 g/mol
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: –17.5 °C
Density: 1.153 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008459
CAS Number: 624-48-6
EINECS: 210-848-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR