1-Bromo-2-methoxyethane

Synonyms: 2-Bromoethyl methyl ether

238155-25G 238155-5G
220046-200EA 78.4 USD
220046-200 220046-645
1-Bromo-2-methoxyethane
1-Bromo-2-methoxyethane
Formula: BrH₂CCH₂OCH₃
MW: 138.99 g/mol
Boiling Pt: 110 °C (1013 hPa)
Density: 1.46 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000236
CAS Number: 6482-24-2
EINECS: 229-339-4
UN: 3271
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR