1,2-Diiodobenzene ≥98%

Warning

238112-1G 238112-10G
220046-196EA 26.92 USD
220046-196 220046-195
1,2-Diiodobenzene ≥98%
1,2-Diiodobenzene
Formula: C₆H₄I₂
MW: 329.91 g/mol
Boiling Pt: 285 °C (1013 hPa)
Density: 2.76 g/cm³ (0 °C)
MDL Number: MFCD00001038
CAS Number: 615-42-9
EINECS: 210-425-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR