(1-Bromoethyl)benzene ≥97%

Warning

Synonyms: 1-Bromo-1-phenylethane, α-Methylbenzyl bromide

238104-100G 238104-25G
220046-192EA 190.62 USD
220046-192 220046-193
(1-Bromoethyl)benzene ≥97%
(1-Bromoethyl)benzene
Formula: C₆H₅CHBrCH₃
MW: 185.06 g/mol
Boiling Pt: 220 °C (1013 hPa)
Density: 0.988 g/cm³ (20 °C)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00000139
CAS Number: 585-71-7
EINECS: 209-560-2
UN: 3334
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f329f7855ae4dffcd097f49d6d504b1ec165e692&vid=6d612184698aeb1969948bdeb536d2ee39ec9551&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR