Yttrium (III) nitrate hexahydrate

Warning

Synonyms: Yttrium trinitrate hexahydrate

237957-100G 237957-500G 237957-25G
220046-622EA 244.62 USD
220046-622 220046-624 220046-623
Yttrium (III) nitrate hexahydrate
Yttrium (III) nitrate hexahydrate
Formula: N₃O₉Y·6H₂O
MW: 383.01 g/mol
Melting Pt: 51.8 °C
Density: 2.682 g/cm³ (20 °C)
MDL Number: MFCD00149940
CAS Number: 13494-98-9
EINECS: 233-802-6
UN: 1477
ADR: 5.1,III

Order Now

Specification Test Results

Assay 99.8% trace metals basis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR