(S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol

Synonyms: (S)-(+)-1,2-O-Isopropylidenglycerol, (S)-(+)-2,2-Dimethyl-1,3-dioxolane-4-methanol, (S)-(+)-1,2-Isopropylideneglycerol, (+)-Solketal, (S)-(+)-2,3-O-Isopropylideneglycerol

237744-1G 237744-5G
220046-147EA 107.69 USD
220046-147 220046-609
(S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol
(S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol
Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Boiling Pt: 82 °C (1013 hPa)
Density: 1.0704 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00063239
CAS Number: 22323-82-6
EINECS: 244-910-8
Merck Index: 12,05232

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR