(S)-(+)-2-Butanol ≥99%

Danger

Synonyms: (S)-(+)-sec-Butyl alcohol, (S)-Butan-2-ol, (S)-(+)-sec-Butanol, (S)-(+)-Ethyl methyl carbinol, (S)-(+)-2-Hydroxy butane, (S)-(+)-Methyl ethyl carbinol

237698-10G 237698-1G
220046-142EA 459.85 USD
220046-142 220046-143
(S)-(+)-2-Butanol ≥99%
(S)-(+)-2-Butanol
Formula: C₄H₁₀O
MW: 74.12 g/mol
Boiling Pt: 99 °C (1013 hPa)
Melting Pt: -114 °C
Density: 0.808 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064281
CAS Number: 4221-99-2
EINECS: 224-168-1
UN: 1120
ADR: 3,III
Merck Index: 13,01540

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR