(2R,3R)-(-)-2,3-Butanediol ≥97%

Synonyms: D(-)-2,3-Butanediol, (2R,3R)-Butane-2,3-diol, (R,R)-(-)-2,3-Butylene glykol, (R,R)-(-)-2,3-Butanediol, (2R,3R)-(-)-2,3-Dihydroxy butane, (R,R)-(-)-2,3-Dihydroxy butane, (2R,3R)-(-)-2,3-Butylene glykol

237639-1G 237639-5G
220045-717EA 77.97 USD
220045-717 220045-718
(2R,3R)-(-)-2,3-Butanediol ≥97%
(2R,3R)-(-)-2,3-Butanediol
Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Boiling Pt: 175 °C (1013 hPa)
Melting Pt: 16 °C
Density: 0.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064267
CAS Number: 24347-58-8
EINECS: 246-186-9
Merck Index: 13,01567

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR