(S)-(-)-1-(1-Naphthyl)ethylamine

Synonyms: (S)-(-)-alpha-(1-Naphthyl)ethylamine

237450-1G 237450-5G
220046-597EA 63.11 USD
220046-597 220046-598
(S)-(-)-1-(1-Naphthyl)ethylamine
(S)-(-)-1-(1-Naphthyl)ethylamine
Formula: C₁₂H₁₃N
MW: 171.24 g/mol
MDL Number: MFCD00064179
CAS Number: 10420-89-0

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR