(R)-(+)-1-(1-Naphthyl)ethylamine

Synonyms: (R)-(+)-α-(1-Naphthyl)ethylamine, (R)-1-(Naphthalen-1-yl)ethanamine, (+)-1-(1-Naphthyl)ethylamine

237442-5G 237442-1G
220046-596EA 152.92 USD
220046-596 220046-595
(R)-(+)-1-(1-Naphthyl)ethylamine
(R)-(+)-1-(1-Naphthyl)ethylamine
Formula: C₁₂H₁₃N
MW: 171.24 g/mol
MDL Number: MFCD00064114
CAS Number: 3886-70-2
EINECS: 223-425-5

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR