1-Chloro-2,4-dinitrobenzene ≥99%

Danger

237329-10G 237329-50G
220046-586EA 40.82 USD
220046-586 220046-587
1-Chloro-2,4-dinitrobenzene ≥99%
1-Chloro-2,4-dinitrobenzene
Formula: C₆H₃ClN₂O₄
MW: 202.55 g/mol
Boiling Pt: 315 °C (1013 hPa)
Melting Pt: 52 - 54 °C
Density: 1.7 g/cm³ (20 °C)
Flash Pt: 194 °C
Storage Temperature: Ambient
MDL Number: MFCD00007075
CAS Number: 97-00-7
EINECS: 202-551-4
UN: 3441
ADR: 6.1,II
Merck Index: 13,02155

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR