Sodium nitrite ≥97.0% ACS

Danger

Synonyms: Diazotizing salts

237213-2.5KG 237213-5G 237213-100G 237213-500G
220046-581EA 185.23 USD
220046-581 220046-583 220046-580 220046-582
Sodium nitrite ≥97.0% ACS
Sodium nitrite
Formula: NaNO₂
MW: 69 g/mol
Boiling Pt: 320 °C (1013 hPa)
Melting Pt: 284 °C
Density: 2.168 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011118
CAS Number: 7632-00-0
EINECS: 231-555-9
UN: 1500
ADR: 5.1,III
Merck Index: 13,08721

Order Now

Specification Test Results

Assay 97.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR