Potassium hydrogen carbonate ≥99.7% ACS

237205-500G 237205-100G 237205-2.5KG
220046-579EA 47.94 USD
220046-579 220046-577 220046-578
Potassium hydrogen carbonate ≥99.7% ACS
Potassium hydrogen carbonate
Formula: KHCO₃
Melting Pt: 292 °C
Density: 2.17 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011402
CAS Number: 298-14-6
EINECS: 206-059-0
Merck Index: 13,07691

Order Now

Specification Test Results

Assay 99.7%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR