Magnesium nitrate hexahydrate ≥99% ACS

Warning

237175-100G 237175-1KG 237175-500G
220046-573EA 28.43 USD
220046-573 220046-575 220046-574 220046-576
Magnesium nitrate hexahydrate ≥99% ACS
Magnesium nitrate hexahydrate
Formula: Mg(NO₃)₂·6H₂O
MW: 256.41 g/mol
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: 89 °C
Density: 1.63 g/cm³ (20 °C)
Flash Pt: 93.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00149779
CAS Number: 13446-18-9
EINECS: 233-826-7
UN: 1474
ADR: 5.1,III
Merck Index: 13,05697

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR