Lead(IV) oxide ≥97.0% ACS

Danger

Synonyms: Lead dioxide

237140-500G 237140-100G
220046-570EA 66.92 USD
220046-570 220046-569
Lead(IV) oxide ≥97.0% ACS
Lead(IV) oxide
Formula: O₂Pb
MW: 239.2 g/mol
Melting Pt: 290 °C
Density: 9.4 g/cm³ (20 °C)
MDL Number: MFCD00011165
CAS Number: 1309-60-0
EINECS: 215-174-5
UN: 1872
ADR: 5.1,III
Merck Index: 12,05424

Order Now

Specification Test Results

Assay 97.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR