Aluminium potassium sulfate dodecahydrate ≥98% ACS

Synonyms: Aluminium Potassium sulphate, 12-Hydrate, Aluminum potassium sulphate dodecahydrate, Aluminium potassium bis(sulphate) dodecahydrate, Potassium alum, Alum potassium

237086-100G 237086-500G 237086-2.5KG
220046-083EA 33.42 USD
220046-083 220046-085 220046-084
Aluminium potassium sulfate dodecahydrate ≥98% ACS
Aluminium potassium sulphate dodecahydrate
Formula: AlK(SO₄)₂·12H₂O
MW: 474.39 g/mol
Melting Pt: 92.5 °C
Density: 1.72 g/cm³ (20 °C)
MDL Number: MFCD00149143
CAS Number: 7784-24-9
EINECS: 233-141-3
Merck Index: 12,00373

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR