Potassium acetate ≥99.0% ACS

Synonyms: Acetic acid potassium salt

236497-2.5KG 236497-100G 236497-500G
220046-547EA 214.77 USD
220046-547 220046-546 220046-548
Potassium acetate ≥99.0% ACS
Potassium acetate
Formula: CH₃COOK
MW: 98.14 g/mol
Melting Pt: 304 °C
Density: 1.57 g/cm³ (25 °C)
Flash Pt: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD00012458
CAS Number: 127-08-2
EINECS: 204-822-2
Merck Index: 13,07687
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13cc4439b794fe60df11a635ce6d3a03e9609099&vid=06c778a684c28fe03ba0262abdbaa81c094e79a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR