Sebacoyl dichloride ≥99%

Danger

Synonyms: Sebacoyl chloride

236365-10G 236365-50G
220046-534EA 117.17 USD
220046-534 220046-535
Sebacoyl dichloride ≥99%
Sebacoyl dichloride
Formula: C₁₀H₁₆Cl₂O₂
MW: 239.14 g/mol
Boiling Pt: 220 °C (1013 hPa)
Melting Pt: –5 °C
Density: 1.12 g/cm³ (20 °C)
Flash Pt: 165 °C
Storage Temperature: Ambient
MDL Number: MFCD00000770
CAS Number: 111-19-3
EINECS: 203-843-4
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR