N-Methylaniline ≥99%

Danger

236233-10G 236233-50G
220046-520EA 120.62 USD
220046-520 220046-521
N-Methylaniline ≥99%
N-Methylaniline
Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 196.3 °C (1013 hPa)
Melting Pt: -57 °C
Density: 0.989 g/cm³ (20 °C)
Flash Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD00008283
CAS Number: 100-61-8
EINECS: 202-870-9
UN: 2294
ADR: 6.1,III
Merck Index: 12,06098

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR