Cobalt(III) fluoride

Danger

Synonyms: Cobaltic fluoride, Cobalt trifluoride

236136-25G 236136-5G
220046-517EA 161.54 USD
220046-517 220046-518
Cobalt(III) fluoride
Cobalt(III) fluoride
Formula: CoF₃
MW: 115.93 g/mol
Melting Pt: 927 °C
Density: 3.88 g/cm³ (20 °C)
MDL Number: MFCD00010942
CAS Number: 10026-18-3
EINECS: 233-062-4
UN: 3260
ADR: 8,II
Merck Index: 12,02491

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR