1,1,1-Trifluoroacetylacetone ≥98%

Danger

Synonyms: 1,1,1-Trifluoropentane-2,4-dione, 1,1,1-Trifluoro-2,4-pentanedione

235970-10G 235970-25G
220046-502EA 88.31 USD
220046-502 220046-503
1,1,1-Trifluoroacetylacetone ≥98%
1,1,1-Trifluoroacetylacetone
Formula: C₅H₅F₃O₂
MW: 154.09 g/mol
Boiling Pt: 107 °C (1013 hPa)
Density: 1.28 g/cm³ (20 °C)
Flash Pt: 28 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000427
CAS Number: 367-57-7
EINECS: 206-698-5
UN: 1224
ADR: 3,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR