Tetramethylammonium iodide

235946-100G 235946-25G
220046-498EA 75.46 USD
220046-498 220046-499
Tetramethylammonium iodide
Tetramethylammonium iodide
Formula: C₄H₁₂IN
MW: 201.05 g/mol
Melting Pt: 300 °C
Density: 1.84 g/cm³ (20 °C)
MDL Number: MFCD00011629
CAS Number: 75-58-1
EINECS: 200-881-3
UN: 2811
ADR: 6.1,III
Merck Index: 12,09364

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR