Butyl methacrylate ≥99%

Synonyms: Methacrylic acid butyl ester

235865-18L 235865-100ML 235865-1L
220046-491EA 639.46 USD
220046-491 220046-490 220046-492
Butyl methacrylate ≥99%
Butyl methacrylate
Formula: C₈H₁₄O₂
MW: 142.2 g/mol
Boiling Pt: 163 °C (1013 hPa)
Melting Pt: –75 °C
Density: 0.8951 g/cm³ (20 °C)
MDL Number: MFCD00009444
CAS Number: 97-88-1
EINECS: 202-615-1
UN: 2227
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR